Product Details

SNP ID
rs9971728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4491902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTATTGTGATTCTCAAACTTTGCG[A/G]GTCTCAGGATCTTTTGCACTCTTAA
Phenotype
MIM: 616082
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf4 PubMed Links

Gene Details

Gene
C12orf4
Gene Name
chromosome 12 open reading frame 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304811.1 Intron NP_001291740.1
NM_020374.3 Intron NP_065107.1
XM_005253716.1 Intron XP_005253773.1
XM_006718992.3 Intron XP_006719055.1

View Full Product Details