Product Details

SNP ID
rs10497344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168875079 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAATATTCATGTGTTTTAGCCTA[C/G]TAATTCCTCTCTGGGAAACAATTCT
Phenotype
MIM: 607496 MIM: 609395
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOSTRIN PubMed Links

Gene Details

Gene
NOSTRIN
Gene Name
nitric oxide synthase trafficking
There are no transcripts associated with this gene.

Gene
SPC25
Gene Name
SPC25, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020675.3 Intron NP_065726.1
XM_011511516.2 Intron XP_011509818.1

View Full Product Details