Product Details

SNP ID
rs5754009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32359006 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCTCATCCATGCGCCATGGGCT[C/T]CAACAGTGGTCCTGGCTACAGAAAA
Phenotype
MIM: 605970 MIM: 605971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFPL3 PubMed Links

Gene Details

Gene
RFPL3
Gene Name
ret finger protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098535.1 Intron NP_001092005.1
NM_006604.2 Intron NP_006595.1
XM_017028534.1 Intron XP_016884023.1
Gene
RFPL3S
Gene Name
RFPL3 antisense
There are no transcripts associated with this gene.

Gene
SLC5A4
Gene Name
solute carrier family 5 member 4
There are no transcripts associated with this gene.

View Full Product Details