Product Details

SNP ID
rs9817063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114128261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAAAGAAATGACCAAGAGGAA[C/T]GGGCCCGTAAAAGTACAAGGTTAGC
Phenotype
MIM: 126451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRD3 PubMed Links

Gene Details

Gene
DRD3
Gene Name
dopamine receptor D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000796.5 1909 Intron NP_000787.2
NM_001282563.2 1909 Intron NP_001269492.1
NM_001290809.1 1909 Intron NP_001277738.1
NM_033663.5 1909 Intron NP_387512.3
XM_017005829.1 1909 UTR 3 XP_016861318.1

View Full Product Details