Product Details

SNP ID
rs10423596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58335780 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTTGGGGGTGACGGAAATACCA[C/G]ATGTCCCTGTGTACCTTCTCTATGC
Phenotype
MIM: 165260
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6806 PubMed Links

Gene Details

Gene
MIR6806
Gene Name
microRNA 6806
There are no transcripts associated with this gene.

Gene
ZSCAN22
Gene Name
zinc finger and SCAN domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321116.1 Intron NP_001308045.1
NM_001321117.1 Intron NP_001308046.1
NM_181846.2 Intron NP_862829.1
XM_006723192.3 Intron XP_006723255.1
XM_011526917.2 Intron XP_011525219.1
XM_011526918.2 Intron XP_011525220.1
XM_017026746.1 Intron XP_016882235.1
XM_017026747.1 Intron XP_016882236.1

View Full Product Details