Product Details
- SNP ID
-
rs10229502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:75072417 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCTTATTGCTGTTAGCAGTAGTA[C/T]AAGGCAGATGAGAAAGTGTGGCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RCC1L
PubMed Links
Gene Details
- Gene
- RCC1L
- Gene Name
- RCC1 like
View Full Product Details