Product Details

SNP ID
rs17882687
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.10:94762760 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCTCTCATGTTTGCTTCTCCTTTCA[A/C]TCTGGAGACAGAGCTCTGGGAGAGG
Phenotype
MIM: 124020
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
CYP2C19*15,c.55A>C CYP2C19*15,g.55A>C
Literature Links
CYP2C19 PubMed Links

Gene Details

Gene
CYP2C19
Gene Name
cytochrome P450 family 2 subfamily C member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000769.2 80 Missense Mutation ATC,CTC I19L NP_000760.1

View Full Product Details