Product Details

SNP ID
rs11124234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238919307 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAATTTATGGAAAATAATTACACC[A/G]TAGCAGAGAGATTTTAATGGTCGTT
Phenotype
MIM: 607556
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ43879 PubMed Links

Gene Details

Gene
FLJ43879
Gene Name
FLJ43879 protein
There are no transcripts associated with this gene.

Gene
TWIST2
Gene Name
twist family bHLH transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details