Product Details

SNP ID
rs7492842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105423090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCAGGAATCGGTGCCTGCCAGC[C/T]GCCTGCAGCTTGCTCATGTGGGGCT
Phenotype
MIM: 603526
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507437 PubMed Links

Gene Details

Gene
LOC100507437
Gene Name
uncharacterized LOC100507437
There are no transcripts associated with this gene.

Gene
MTA1
Gene Name
metastasis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203258.1 Intron NP_001190187.1
NM_004689.3 Intron NP_004680.2
XM_011537305.1 Intron XP_011535607.1
XM_011537306.1 Intron XP_011535608.1
XM_011537307.1 Intron XP_011535609.1
XM_011537308.2 Intron XP_011535610.1
XM_011537309.1 Intron XP_011535611.1
XM_011537310.1 Intron XP_011535612.1
XM_017021752.1 Intron XP_016877241.1
XM_017021753.1 Intron XP_016877242.1
XM_017021754.1 Intron XP_016877243.1
XM_017021755.1 Intron XP_016877244.1
XM_017021756.1 Intron XP_016877245.1
XM_017021757.1 Intron XP_016877246.1
XM_017021758.1 Intron XP_016877247.1
XM_017021759.1 Intron XP_016877248.1
Gene
TEX22
Gene Name
testis expressed 22
There are no transcripts associated with this gene.

View Full Product Details