Product Details

SNP ID
rs6468325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30408686 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCACTGTGTTCTGGTGACAGTAA[C/G]ACCCAATGTTGAGAAATAACTCTCT
Phenotype
MIM: 601558
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBPMS PubMed Links

Gene Details

Gene
RBPMS
Gene Name
RNA binding protein with multiple splicing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008710.2 Intron NP_001008710.1
NM_001008711.2 Intron NP_001008711.1
NM_001008712.2 Intron NP_001008712.1
NM_006867.3 Intron NP_006858.1
XM_017012980.1 Intron XP_016868469.1
XM_017012981.1 Intron XP_016868470.1
XM_017012982.1 Intron XP_016868471.1
XM_017012983.1 Intron XP_016868472.1
XM_017012984.1 Intron XP_016868473.1
XM_017012985.1 Intron XP_016868474.1
XM_017012986.1 Intron XP_016868475.1
XM_017012987.1 Intron XP_016868476.1
XM_017012988.1 Intron XP_016868477.1
XM_017012989.1 Intron XP_016868478.1
XM_017012990.1 Intron XP_016868479.1
XM_017012991.1 Intron XP_016868480.1
XM_017012992.1 Intron XP_016868481.1

View Full Product Details