Product Details

SNP ID
rs12788491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77820135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAGTAAGGGTGGGGAGGAGGA[G/T]AAAAGCGCAAAGCTGGCCGGAACCT
Phenotype
MIM: 608522
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AAMDC PubMed Links

Gene Details

Gene
AAMDC
Gene Name
adipogenesis associated Mth938 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316957.1 Intron NP_001303886.1
NM_001316958.1 Intron NP_001303887.1
NM_001316960.1 Intron NP_001303889.1
NM_001316961.1 Intron NP_001303890.1
NM_001316962.1 Intron NP_001303891.1
NM_024684.3 Intron NP_078960.1
XM_011544968.2 Intron XP_011543270.1
XM_011544969.2 Intron XP_011543271.1
XM_017017620.1 Intron XP_016873109.1
Gene
RPS20P27
Gene Name
ribosomal protein S20 pseudogene 27
There are no transcripts associated with this gene.

Gene
RSF1
Gene Name
remodeling and spacing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016578.3 Intron NP_057662.3
XM_005274051.2 Intron XP_005274108.1
XM_017017923.1 Intron XP_016873412.1

View Full Product Details