Product Details

SNP ID
rs11621365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105405432 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATTTAATTCAGTCAATGAAAAC[A/G]GACTAAGTGTTCCAGTTAAAAGACA
Phenotype
MIM: 610423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PACS2 PubMed Links

Gene Details

Gene
PACS2
Gene Name
phosphofurin acidic cluster sorting protein 2
There are no transcripts associated with this gene.

Gene
TEX22
Gene Name
testis expressed 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195082.1 Intron NP_001182011.1
XM_006720234.3 Intron XP_006720297.1

View Full Product Details