Product Details

SNP ID
rs4965537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99718911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGAAATACATGTAGTTATGCGC[A/G]CACACACACACACACACACACACAC
Phenotype
MIM: 600660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYSMD4 PubMed Links

Gene Details

Gene
LYSMD4
Gene Name
LysM domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284417.1 Intron NP_001271346.1
NM_001284418.1 Intron NP_001271347.1
NM_001284419.1 Intron NP_001271348.1
NM_001284420.1 Intron NP_001271349.1
NM_001284421.1 Intron NP_001271350.1
NM_001284422.1 Intron NP_001271351.1
NM_152449.3 Intron NP_689662.2
XM_006720397.3 Intron XP_006720460.2
XM_011521241.1 Intron XP_011519543.1
XM_011521242.2 Intron XP_011519544.1
XM_011521243.2 Intron XP_011519545.1
XM_011521244.1 Intron XP_011519546.1
XM_011521245.2 Intron XP_011519547.1
XM_017021926.1 Intron XP_016877415.1
XM_017021927.1 Intron XP_016877416.1
XM_017021928.1 Intron XP_016877417.1
XM_017021929.1 Intron XP_016877418.1
XM_017021930.1 Intron XP_016877419.1
Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
There are no transcripts associated with this gene.

View Full Product Details