Product Details
- SNP ID
-
rs10784411
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:64462147 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCCCCCATCCTAAATCATGCTGT[G/T]AGACTCTCCAGTGGCCAAAGCCCCC
- Phenotype
-
MIM: 604834
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBK1
PubMed Links
Gene Details
- Gene
- TBK1
- Gene Name
- TANK binding kinase 1
View Full Product Details