Product Details

SNP ID
hCV30719588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71729135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCATTTTATTCCTCACTCGCCT[C/T]TAAGACTTATCATTTAACTTTAAAT
Phenotype
MIM: 603533 MIM: 611066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1G1 PubMed Links

Gene Details

Gene
AP1G1
Gene Name
adaptor related protein complex 1 gamma 1 subunit
There are no transcripts associated with this gene.

Gene
PHLPP2
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
There are no transcripts associated with this gene.

View Full Product Details