Product Details

SNP ID
rs11706359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:155870359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGATTCGGCTTCGGTTTTTCTCC[C/T]GGCCGGCGCCACAGGCCCCTCCTCC
Phenotype
MIM: 600358
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMPS PubMed Links

Gene Details

Gene
GMPS
Gene Name
guanine monophosphate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003875.2 Intron NP_003866.1
XM_011513263.2 Intron XP_011511565.1
XM_017007422.1 Intron XP_016862911.1

View Full Product Details