Product Details
- SNP ID
-
rs12582136
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49625584 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCTAAGTTCTGAGATGCCCCTTA[C/G]ATGTGAAGTCCCAGAAGTATTGATT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRPF40B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77938251] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRPF40B
- Gene Name
- pre-mRNA processing factor 40 homolog B
View Full Product Details