Product Details

SNP ID
rs12049882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27515789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCATGTAACTTATCTAATTTGT[G/T]GAAAAATATATTTGTCACACAATTC
Phenotype
MIM: 611468 MIM: 612332
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BDNF-AS PubMed Links

Gene Details

Gene
BDNF-AS
Gene Name
BDNF antisense RNA
There are no transcripts associated with this gene.

Gene
LIN7C
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
There are no transcripts associated with this gene.

Gene
MIR8087
Gene Name
microRNA 8087
There are no transcripts associated with this gene.

View Full Product Details