Product Details

SNP ID
rs13086084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44338996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCCGCATCGCATCTACATTTTA[A/C]CTCGCTGTGAAATGAGAACGCAAAG
Phenotype
MIM: 614412
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TCAIM PubMed Links

Gene Details

Gene
TCAIM
Gene Name
T-cell activation inhibitor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029839.2 Intron NP_001025010.1
NM_001029840.2 Intron NP_001025011.1
NM_001282913.1 Intron NP_001269842.1
NM_001282914.1 Intron NP_001269843.1
NM_001282915.1 Intron NP_001269844.1
NM_173826.3 Intron NP_776187.2
XM_017006251.1 Intron XP_016861740.1
Gene
TOPAZ1
Gene Name
testis and ovary specific PAZ domain containing 1
There are no transcripts associated with this gene.

View Full Product Details