Product Details

SNP ID
rs12086853
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:155129086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGGTGAGGCAGTCTGTTGTCTG[C/T]GGCTTGTTAGGGTGGGGGAGAGAAC
Phenotype
MIM: 191164 MIM: 613683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFNA1 PubMed Links

Gene Details

Gene
EFNA1
Gene Name
ephrin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004428.2 Intron NP_004419.2
NM_182685.1 Intron NP_872626.1
Gene
SLC50A1
Gene Name
solute carrier family 50 member 1
There are no transcripts associated with this gene.

View Full Product Details