Product Details
- SNP ID
-
rs12058480
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:155128369 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGCCTTCAGCATAGACTTCGCCC[C/T]TGGCAGCCAGGGCCCTTGTTCCAGG
- Phenotype
-
MIM: 191164
MIM: 613683
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFNA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76547070] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFNA1
- Gene Name
- ephrin A1
- Gene
- SLC50A1
- Gene Name
- solute carrier family 50 member 1
There are no transcripts associated with this gene.
View Full Product Details