Product Details

SNP ID
rs11168373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48047810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTTAGTGCATTAGGATAGTAA[C/T]TGCTCCCATTAGGGCATACTTTGGT
Phenotype
MIM: 612157
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SENP1 PubMed Links

Gene Details

Gene
SENP1
Gene Name
SUMO1/sentrin specific peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267594.1 Intron NP_001254523.1
NM_001267595.1 Intron NP_001254524.1
XM_006719361.3 Intron XP_006719424.1
XM_006719362.2 Intron XP_006719425.1
XM_011538244.2 Intron XP_011536546.1
XM_011538245.2 Intron XP_011536547.1
XM_017019229.1 Intron XP_016874718.1
XM_017019230.1 Intron XP_016874719.1
XM_017019231.1 Intron XP_016874720.1
XM_017019232.1 Intron XP_016874721.1
XM_017019233.1 Intron XP_016874722.1
XM_017019234.1 Intron XP_016874723.1
XM_017019235.1 Intron XP_016874724.1
XM_017019236.1 Intron XP_016874725.1
XM_017019237.1 Intron XP_016874726.1
XM_017019238.1 Intron XP_016874727.1
XM_017019239.1 Intron XP_016874728.1
XM_017019240.1 Intron XP_016874729.1

View Full Product Details