Product Details

SNP ID
rs11633560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79416006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTCTTGTTCATATGCCAAGGTA[A/G]ATATTTTCTTTGTTGCAATGGTAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1024 PubMed Links

Gene Details

Gene
KIAA1024
Gene Name
KIAA1024
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015206.2 5212 Intron NP_056021.1
XM_011521392.1 5212 Intron XP_011519694.1
XM_011521393.2 5212 Intron XP_011519695.1
XM_011521395.2 5212 Intron XP_011519697.1
XM_011521396.2 5212 Intron XP_011519698.1
XM_017022027.1 5212 Intron XP_016877516.1
Gene
TMED3
Gene Name
transmembrane p24 trafficking protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301203.1 5212 Intron NP_001288132.1
NM_007364.3 5212 Intron NP_031390.1
XM_017022040.1 5212 UTR 3 XP_016877529.1
XM_017022041.1 5212 Intron XP_016877530.1

View Full Product Details