Product Details

SNP ID
rs12909104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79421561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGTCAAGCTCCAATTGAACAC[C/T]TCTTGGATTTGAATGAAGTTCTAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1024 PubMed Links

Gene Details

Gene
KIAA1024
Gene Name
KIAA1024
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015206.2 Intron NP_056021.1
XM_011521392.1 Intron XP_011519694.1
XM_011521393.2 Intron XP_011519695.1
XM_011521395.2 Intron XP_011519697.1
XM_011521396.2 Intron XP_011519698.1
XM_017022027.1 Intron XP_016877516.1
Gene
TMED3
Gene Name
transmembrane p24 trafficking protein 3
There are no transcripts associated with this gene.

View Full Product Details