Product Details
- SNP ID
-
rs4886987
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78002045 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACACTTCTAGTCAATAAAGCATAA[A/G]GAACAAACATAAGGAATCATTTTAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC91450
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79991040] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC91450
- Gene Name
- uncharacterized LOC91450
There are no transcripts associated with this gene.
- Gene
- TBC1D2B
- Gene Name
- TBC1 domain family member 2B
View Full Product Details