Product Details

SNP ID
rs12010853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:35799475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCCCTGGGAGACTCACCCTA[C/G]GGGTGTCAAACTGAAGATACTGCTA
Phenotype
MIM: 300762
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAGEB16 PubMed Links

Gene Details

Gene
MAGEB16
Gene Name
MAGE family member B16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099921.1 Intron NP_001093391.1
XM_011545453.2 Intron XP_011543755.1
XM_011545454.2 Intron XP_011543756.1

View Full Product Details