Product Details

SNP ID
rs13018579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200490383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGGTGCCCCCGTGCCCACCAGG[A/G]CCGTGGCTCTGGAAGGTGCAGAGCG
Phenotype
MIM: 613817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD18 PubMed Links

Gene Details

Gene
KCTD18
Gene Name
potassium channel tetramerization domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321547.1 1540 Missense Mutation GCC,GTC A333V NP_001308476.1
NM_001321548.1 1540 Missense Mutation GCC,GTC A124V NP_001308477.1
NM_001321550.1 1540 Missense Mutation GCC,GTC A124V NP_001308479.1
NM_152387.3 1540 Missense Mutation GCC,GTC A333V NP_689600.2
Gene
SPATS2L
Gene Name
spermatogenesis associated serine rich 2 like
There are no transcripts associated with this gene.

View Full Product Details