Product Details

SNP ID
rs12296916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:90952736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGATAAAGACTATATCCAATAA[A/C]GAGGCTTCAAAACCTAAGCCATGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCER1 PubMed Links

Gene Details

Gene
CCER1
Gene Name
coiled-coil glutamate rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152638.3 2441 UTR 3 NP_689851.1
Gene
LINC00615
Gene Name
long intergenic non-protein coding RNA 615
There are no transcripts associated with this gene.

View Full Product Details