Product Details
- SNP ID
-
rs11105882
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:90953885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACCTCCTGGTCATACTCCTCCTC[A/C]TCATCATTTTTCTTCTCCTCCTCTT
- Phenotype
-
MIM: 601657
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCER1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78240650] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCER1
- Gene Name
- coiled-coil glutamate rich protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152638.3 |
1292 |
Missense Mutation |
GAG,GAT |
E286D |
NP_689851.1 |
- Gene
- EPYC
- Gene Name
- epiphycan
There are no transcripts associated with this gene.
- Gene
- LINC00615
- Gene Name
- long intergenic non-protein coding RNA 615
There are no transcripts associated with this gene.
View Full Product Details