Product Details

SNP ID
rs12814406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48935519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCCCTTTCTTTGCCTGGAGCC[A/G]GACACTGGTAGCAGGGGAGGAACAG
Phenotype
MIM: 103190 MIM: 610571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARF3 PubMed Links

Gene Details

Gene
ARF3
Gene Name
ADP ribosylation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001659.2 Intron NP_001650.1
XM_005268856.1 Intron XP_005268913.1
XM_006719391.3 Intron XP_006719454.1
Gene
FKBP11
Gene Name
FK506 binding protein 11
There are no transcripts associated with this gene.

View Full Product Details