Product Details

SNP ID
rs12409537
Assay Type
Functionally Tested
NCBI dbSNP Submissions
18
Location
Chr.1:151219024 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAAAATTAGACTAAAAATCTCTT[C/T]CTGAATGATTTTGTTCTAATATAAG
Phenotype
MIM: 603275
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PIP5K1A PubMed Links

Gene Details

Gene
PIP5K1A
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135636.1 Intron NP_001129108.1
NM_001135637.1 Intron NP_001129109.1
NM_001135638.1 Intron NP_001129110.1
NM_003557.2 Intron NP_003548.1
XM_005245525.4 Intron XP_005245582.1
XM_005245527.3 Intron XP_005245584.1
XM_005245528.4 Intron XP_005245585.1
XM_005245530.4 Intron XP_005245587.1
XM_005245531.3 Intron XP_005245588.1
XM_005245532.3 Intron XP_005245589.1
XM_006711563.3 Intron XP_006711626.1
XM_006711564.3 Intron XP_006711627.1
XM_006711567.3 Intron XP_006711630.1
XM_006711568.3 Intron XP_006711631.2
XM_011510043.2 Intron XP_011508345.1
XM_011510045.2 Intron XP_011508347.1
XM_011510046.2 Intron XP_011508348.1
XM_011510048.2 Intron XP_011508350.1
XM_011510049.2 Intron XP_011508351.1
XM_017002439.1 Intron XP_016857928.1
XM_017002440.1 Intron XP_016857929.1
XM_017002441.1 Intron XP_016857930.1

View Full Product Details