Product Details

SNP ID
rs12251120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:61409390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACTTCCAGTAGTCGCGGTTCAA[A/C]GAAGCTGCAGCTTTGTTCACTACTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM26 PubMed Links

Gene Details

Gene
TMEM26
Gene Name
transmembrane protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178505.7 2408 UTR 3 NP_848600.2
XM_011539450.2 2408 Intron XP_011537752.1
XM_011539451.1 2408 Intron XP_011537753.1
XM_017015829.1 2408 Intron XP_016871318.1

View Full Product Details