Product Details

SNP ID
rs12044203
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:35810798 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTTGTCATATCAGTTGCCCTTT[C/T]AGAGAATGCAGTTATCTGACTTTGT
Phenotype
MIM: 607356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGO4 PubMed Links

Gene Details

Gene
AGO4
Gene Name
argonaute 4, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017629.3 Intron NP_060099.2
XM_005270578.3 Intron XP_005270635.1
XM_005270579.3 Intron XP_005270636.1
XM_011540883.2 Intron XP_011539185.1
XM_011540884.2 Intron XP_011539186.1
XM_017000529.1 Intron XP_016856018.1

View Full Product Details