Product Details

SNP ID
rs11970404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28430509 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGAGGAAAACGGGGAAGATGTTG[G/T]CTTGTTCTCCATTGTTTTAAACCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN23 PubMed Links

Gene Details

Gene
ZSCAN23
Gene Name
zinc finger and SCAN domain containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012455.1 2905 Intron NP_001012458.1
XM_005248950.2 2905 UTR 3 XP_005249007.1
XM_011514406.2 2905 Intron XP_011512708.1
XM_011514408.2 2905 Intron XP_011512710.1
XM_011514410.2 2905 Intron XP_011512712.1
XM_017010479.1 2905 Intron XP_016865968.1

View Full Product Details