Product Details

SNP ID
rs12125833
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:44742976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCTGAGAATGGGCTGGGCTTA[A/G]AGGGTGGAGTGTGAAGGCTTAGCTG
Phenotype
MIM: 604538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF2C PubMed Links

Gene Details

Gene
KIF2C
Gene Name
kinesin family member 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297655.1 Intron NP_001284584.1
NM_001297656.1 Intron NP_001284585.1
NM_001297657.1 Intron NP_001284586.1
NM_006845.3 Intron NP_006836.2
XM_011540540.2 Intron XP_011538842.1
XM_011540541.2 Intron XP_011538843.1

View Full Product Details