Product Details
- SNP ID
-
rs11042814
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:10454315 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTTTCTGTCTGAACCCGATGCA[A/G]TGGGTCTCATACCTGTAAACTGGGC
- Phenotype
-
MIM: 102772
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMPD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75913860] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMPD3
- Gene Name
- adenosine monophosphate deaminase 3
View Full Product Details