Product Details

SNP ID
rs12313051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50420003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTATGAAGGATGTAAGTCATTA[A/G]GATCATGATTTAGCAGAAAAAACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LARP4 PubMed Links

Gene Details

Gene
LARP4
Gene Name
La ribonucleoprotein domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170803.1 Intron NP_001164274.1
NM_001170804.1 Intron NP_001164275.1
NM_001170808.1 Intron NP_001164279.1
NM_052879.4 Intron NP_443111.4
NM_199188.2 Intron NP_954658.2
NM_199190.2 Intron NP_954660.1
XM_005268613.2 Intron XP_005268670.1
XM_011537828.2 Intron XP_011536130.1
XM_011537829.2 Intron XP_011536131.1
XM_011537830.2 Intron XP_011536132.1
XM_011537831.2 Intron XP_011536133.1
XM_011537833.2 Intron XP_011536135.1
XM_011537834.2 Intron XP_011536136.1
XM_011537835.2 Intron XP_011536137.1
XM_011537836.2 Intron XP_011536138.1
XM_011537838.2 Intron XP_011536140.1
XM_011537839.2 Intron XP_011536141.1
XM_011537840.2 Intron XP_011536142.1
XM_011537841.2 Intron XP_011536143.1
XM_011537843.2 Intron XP_011536145.1
XM_011537844.2 Intron XP_011536146.1
XM_017018750.1 Intron XP_016874239.1
XM_017018751.1 Intron XP_016874240.1
XM_017018752.1 Intron XP_016874241.1
XM_017018753.1 Intron XP_016874242.1
XM_017018754.1 Intron XP_016874243.1
XM_017018755.1 Intron XP_016874244.1
XM_017018756.1 Intron XP_016874245.1
XM_017018757.1 Intron XP_016874246.1
XM_017018758.1 Intron XP_016874247.1
XM_017018759.1 Intron XP_016874248.1
XM_017018760.1 Intron XP_016874249.1
XM_017018761.1 Intron XP_016874250.1
XM_017018762.1 Intron XP_016874251.1

View Full Product Details