Product Details

SNP ID
rs10783790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56537494 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGTGTAATGTCCTCGTGGTTCA[C/T]CTATGTTGTAGCTTGTGTCAGTGTT
Phenotype
MIM: 602387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBMS2 PubMed Links

Gene Details

Gene
RBMS2
Gene Name
RNA binding motif single stranded interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002898.3 Intron NP_002889.1
XM_005269059.4 Intron XP_005269116.1
XM_005269060.4 Intron XP_005269117.1
XM_005269061.3 Intron XP_005269118.1
XM_005269066.4 Intron XP_005269123.1
XM_006719541.3 Intron XP_006719604.1
XM_006719542.3 Intron XP_006719605.1
XM_006719543.3 Intron XP_006719606.1
XM_006719544.3 Intron XP_006719607.1
XM_011538637.2 Intron XP_011536939.1
XM_011538638.2 Intron XP_011536940.1
XM_011538639.2 Intron XP_011536941.1
XM_011538640.2 Intron XP_011536942.1
XM_011538642.2 Intron XP_011536944.1
XM_017019775.1 Intron XP_016875264.1
XM_017019776.1 Intron XP_016875265.1
XM_017019777.1 Intron XP_016875266.1
XM_017019778.1 Intron XP_016875267.1

View Full Product Details