Product Details

SNP ID
rs12248366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73170720 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTACATGGTAGCAGTTTAGTTCC[C/T]AGTAAACTCTTTGGGAACTAGGAAG
Phenotype
MIM: 616464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECD PubMed Links

Gene Details

Gene
ECD
Gene Name
ecdysoneless cell cycle regulator
There are no transcripts associated with this gene.

Gene
FAM149B1
Gene Name
family with sequence similarity 149 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173348.1 Intron NP_775483.1
XM_005269744.2 Intron XP_005269801.1
XM_005269745.2 Intron XP_005269802.1
XM_005269747.3 Intron XP_005269804.1
XM_011539737.2 Intron XP_011538039.1
XM_011539740.2 Intron XP_011538042.1
XM_017016164.1 Intron XP_016871653.1
XM_017016165.1 Intron XP_016871654.1
XM_017016166.1 Intron XP_016871655.1
XM_017016167.1 Intron XP_016871656.1

View Full Product Details