Product Details
- SNP ID
-
rs12085387
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:235978302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTATAACATAGAATTGGAATTTTA[A/C]AGTGCAAAGTTCTTTTTATGTAGCT
- Phenotype
-
MIM: 131390
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NID1
PubMed Links
Gene Details
- Gene
- NID1
- Gene Name
- nidogen 1
View Full Product Details