Product Details

SNP ID
rs12253011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70419252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTTGACCATTTGTAAGCAGCAC[G/T]TTTCCATAACTGTTTATCAGTTAAA
Phenotype
MIM: 602224
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF4EBP2 PubMed Links

Gene Details

Gene
EIF4EBP2
Gene Name
eukaryotic translation initiation factor 4E binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004096.4 Intron NP_004087.1

View Full Product Details