Product Details
- SNP ID
-
rs12267561
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:70155558 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCTAGGTGCAGGGGTTTTGCAAA[G/T]AAATAAGACACAGTCCCTATCATGT
- Phenotype
-
MIM: 607691
MIM: 611017
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SAR1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148248029] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SAR1A
- Gene Name
- secretion associated Ras related GTPase 1A
- Gene
- TYSND1
- Gene Name
- trypsin domain containing 1
There are no transcripts associated with this gene.
View Full Product Details