Product Details

SNP ID
rs12856054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:51407623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAACCCAGCCAGGCCCTGCACTC[C/T]TAAGCCACGCATCTGAGGCAAGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXorf67 PubMed Links

Gene Details

Gene
CXorf67
Gene Name
chromosome X open reading frame 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203407.2 709 Silent Mutation CTA,TTA L203L NP_981952.1

View Full Product Details