Product Details
- SNP ID
-
rs12695695
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139347807 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTAACTCATAAGGTAATTGAGGCC[A/G]TTAAATCAGCCAAATACATTTAGAC
- Phenotype
-
MIM: 606990
MIM: 605810
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COPB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80248005] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COPB2
- Gene Name
- coatomer protein complex subunit beta 2
There are no transcripts associated with this gene.
- Gene
- MRPS22
- Gene Name
- mitochondrial ribosomal protein S22
View Full Product Details