Product Details
- SNP ID
-
rs12807715
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:17266945 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAACTTTAAGGCTTGTTTGCTGGA[A/G]CTGGTGGGTGGGAGTGGGGTGGTGT
- Phenotype
-
MIM: 608020
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUCB2
PubMed Links
Gene Details
- Gene
- NUCB2
- Gene Name
- nucleobindin 2
View Full Product Details