Product Details

SNP ID
rs4665057
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159318483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTCTCTTCATGATAAAAAGCA[C/G]TGGACATTATCTAAGGCCCACTTCA
Phenotype
MIM: 605683
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
BAZ2B PubMed Links

Gene Details

Gene
BAZ2B
Gene Name
bromodomain adjacent to zinc finger domain 2B
There are no transcripts associated with this gene.

View Full Product Details