Product Details
- SNP ID
-
rs10981589
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112997239 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGGTCTCTCCCTTGTATGTATTC[A/G]CTTATGTTTAGTTAGAGCTGAGCTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF883
PubMed Links
Gene Details
- Gene
- ZNF883
- Gene Name
- zinc finger protein 883
View Full Product Details