Product Details

SNP ID
rs8058557
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70379581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTTAAATTACTATTGCAGAAAA[T/C]CTGGCTGGATGCTGGGACCTACCTC
Phenotype
MIM: 607188
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
DDX19A PubMed Links

Gene Details

Gene
DDX19A
Gene Name
DEAD-box helicase 19A
There are no transcripts associated with this gene.

Gene
ST3GAL2
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006927.3 4304 UTR 3 NP_008858.1

View Full Product Details