Product Details

SNP ID
rs11860432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31074724 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGGGGCCGCCGGTGCGAGGTA[G/T]GCGTCCTGAGAGGAAATTGGCAGAC
Phenotype
MIM: 610561
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014699.3 952 Intron NP_055514.3
XM_005255710.3 952 UTR 5 XP_005255767.1
XM_005255711.3 952 Intron XP_005255768.1
XM_005255712.3 952 UTR 5 XP_005255769.1
XM_011545990.2 952 Intron XP_011544292.1
Gene
ZNF668
Gene Name
zinc finger protein 668
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172668.1 952 Intron NP_001166139.1
NM_001172669.1 952 Intron NP_001166140.1
NM_001172670.1 952 Intron NP_001166141.1
NM_024706.4 952 Intron NP_078982.3

View Full Product Details