Product Details

SNP ID
rs12048091
Assay Type
Functionally Tested
NCBI dbSNP Submissions
36
Location
Chr.1:154422601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACATCATGGGTGCACAAAAATT[A/G]TTGAATTCTTGAAGAAGGGACAGGC
Phenotype
MIM: 147880
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IL6R PubMed Links
Additional Information
For this assay, SNP(s) [rs75525739] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
IL6R
Gene Name
interleukin 6 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000565.3 Intron NP_000556.1
NM_001206866.1 Intron NP_001193795.1
NM_181359.2 Intron NP_852004.1
XM_005245139.1 Intron XP_005245196.1
XM_005245140.2 Intron XP_005245197.1
XM_006711298.1 Intron XP_006711361.1
XM_006711299.3 Intron XP_006711362.1
XM_017001199.1 Intron XP_016856688.1
XM_017001200.1 Intron XP_016856689.1
XM_017001201.1 Intron XP_016856690.1

View Full Product Details